Learn More
Biomatik Corporation Goat Insulin (INS) ELISA Kit
Supplier:  Biomatik Corporation EKU11003
Encompass_Preferred
Goat Insulin (INS) ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.62pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: A0A452E6C9. Gene ID: 108634241. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3248