Learn More
Biomatik Corporation Mouse Vascular Endothelial Growth Factor A (VEGFA) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11273
Encompass_Preferred
Mouse Vascular Endothelial Growth Factor A (VEGFA) Mini sample ELISA Kit for detecting VEGFA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00731. Gene ID: 22339. Target Synonyms: VEGF; VEGF-A; VPF; Vascular Endothelial Cell Growth Factor A; Vascular Permeability Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3274