Learn More
Biomatik Corporation Human Sestrin 1 (SESN1) ELISA Kit
Supplier:  Biomatik Corporation EKU11425
Encompass_Preferred
Human Sestrin 1 (SESN1) ELISA Kit for detecting SESN1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y6P5. Gene ID: 27244. Target Synonyms: SEST1; PA26; p53-regulated protein PA26. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3290