Learn More
Biomatik Corporation Mouse Mitochondrial Uncoupling Protein 2 (UCP2) ELISA Kit
Supplier:  Biomatik Corporation EKU11507
Encompass_Preferred
Mouse Mitochondrial Uncoupling Protein 2 (UCP2) ELISA Kit for detecting UCP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70406. Gene ID: 22228. Target Synonyms: SLC25A8; UCPH; Solute carrier family 25 member 8 ;Uncoupling Protein 2, Mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3300