Learn More
Biomatik Corporation Vanillylmandelic Acid (VMA) ELISA Kit
Supplier:  Biomatik Corporation EKU11512
Encompass_Preferred
Vanillylmandelic Acid (VMA) ELISA Kit for detecting VMA in urine, biological agents. Species Reactivity: Universal. Detection Range: 6.17-500ng/ml. Sensitivity: 2.69ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Vanillyl Mandelic Acid. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3302