Learn More
Biomatik Corporation Mouse Thyroid Stimulating Hormone (TSH) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11621
Encompass_Preferred
Mouse Thyroid Stimulating Hormone (TSH) Mini sample ELISA Kit for detecting TSH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 49.4-4,000pg/ml. Sensitivity: 16.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01216; P12656. Gene ID: 12640. Target Synonyms: Thyrotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3322