Learn More
Biomatik Corporation Rat Coagulation Factor XII (F12) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11624
Encompass_Preferred
Rat Coagulation Factor XII (F12) Mini sample ELISA Kit for detecting F12 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.232ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZTE0. Gene ID: 306761. Target Synonyms: FXII; HAF; Contact Factor; Hageman Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3324