Learn More
Biomatik Corporation Dog Kidney Injury Molecule 1 (Kim1) ELISA Kit
Supplier:  Biomatik Corporation EKU11680
Encompass_Preferred
Dog Kidney Injury Molecule 1 (Kim1) ELISA Kit for detecting Kim1 in serum, plasma, urine, tissue homogenates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1CME9. Gene ID: 612069. Target Synonyms: HAVCR1; TIM1; TIMD1; HAVCR; Hepatitis A Virus Cellular Receptor 1; T Cell Immunoglobulin And Mucin Domain-Containing Protein 1; T-cell immunoglobulin mucin receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3329