Learn More
Biomatik Corporation Human Relaxin 2 (RLN2) ELISA Kit
Supplier:  Biomatik Corporation EKU11752
Encompass_Preferred
Human Relaxin 2 (RLN2) ELISA Kit for detecting RLN2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04090. Gene ID: 6019. Target Synonyms: H2; RLXH2; Relaxin H2; Prorelaxin H2; Relaxin,Ovarian,Of Pregnancy; Prorelaxin H2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3340