Learn More
Biomatik Corporation Human Cysteine And Glycine Rich Protein 3 (CSRP3) ELISA Kit
Supplier:  Biomatik Corporation EKU11771
Encompass_Preferred
Human Cysteine And Glycine Rich Protein 3 (CSRP3) ELISA Kit for detecting CSRP3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50461. Gene ID: 8048. Target Synonyms: CLP; CRP3; MLP; CMD1M; LMO4; Cardiac LIM Protein; Cysteine-rich protein 3; LIM domain protein, cardiac; Muscle LIM protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3342