Learn More
Biomatik Corporation Human Glycerol-3-Phosphate Dehydrogenase 1 (GPD1) ELISA Kit
Supplier:  Biomatik Corporation EKU11803
Encompass_Preferred
Human Glycerol-3-Phosphate Dehydrogenase 1 (GPD1) ELISA Kit for detecting GPD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21695. Gene ID: 2819. Target Synonyms: GPDH-C; GPD-C; Glycerol-3-phosphate dehydrogenase [NAD (+) ], cytoplasmic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3347