Learn More
Biomatik Corporation Rabbit Collagen Type IV (COL4) ELISA Kit
Supplier:  Biomatik Corporation EKU11877
Encompass_Preferred
Rabbit Collagen Type IV (COL4) ELISA Kit for detecting COL4 in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: Type-IV Collagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3361