Learn More
Biomatik Corporation Rat Insulin (INS) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11890
Encompass_Preferred
Rat Insulin (INS) Mini sample ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 49.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01322. Gene ID: 24505. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3364