Learn More
Biomatik Corporation Rabbit Immunoglobulin A (IgA) ELISA Kit
Supplier:  Biomatik Corporation EKU11902
Encompass_Preferred
Rabbit Immunoglobulin A (IgA) ELISA Kit for detecting IgA in serum, plasma,, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.46ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01879. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3367