Learn More
Biomatik Corporation Dog Secretory Immunoglobulin A (sIgA) ELISA Kit
Supplier:  Biomatik Corporation EKU11920
Encompass_Preferred
Dog Secretory Immunoglobulin A (sIgA) ELISA Kit for detecting sIgA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200ng/ml. Sensitivity: 1.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: S-IgA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3371