Learn More
Biomatik Corporation Rat Gastric Inhibitory Polypeptide (GIP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11941
Encompass_Preferred
Rat Gastric Inhibitory Polypeptide (GIP) Mini sample ELISA Kit for detecting GIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 61.7-5,000pg/ml. Sensitivity: 21.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06145. Gene ID: 25040. Target Synonyms: Incretin hormone; Glucose Dependent Insulinotropic Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3378