Learn More
Biomatik Corporation Mouse Aspartate Aminotransferase (AST) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11950
Encompass_Preferred
Mouse Aspartate Aminotransferase (AST) Mini sample ELISA Kit for detecting AST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05201. Gene ID: 14718. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase, cytoplasmic; Transaminase A; Aspartate Transaminase 1,Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1,Soluble. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3382