Learn More
Biomatik Corporation Human cross linked N-telopeptide of type Ⅰ collagen, NTX ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33310
Encompass_Preferred
The Human cross linked N-telopeptide of type Ⅰ collagen, NTX ELISA Kit allows for quantitative determination of cross linked N-telopeptide of type Ⅰ collagen, NTX in serum, plasma and urine. Species Reactivity: Human. Detection Range: 7.8 nM BCE-500 nM BCE. Sensitivity: 1.95 nM BCE. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7812