Learn More
Biomatik Corporation Human Neuronal Cell Adhesion Molecule (NRCAM) ELISA Kit
Supplier:  Biomatik Corporation EKU12004
Encompass_Preferred
Human Neuronal Cell Adhesion Molecule (NRCAM) ELISA Kit for detecting NRCAM in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92823. Gene ID: 4897. Target Synonyms: Bravo; NgCAM-Related Cell Adhesion Molecule; Neuronal surface protein Bravo. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3397