Learn More
Biomatik Corporation Human Kruppel Like Factor 15 (KLF15) CLIA Kit
Supplier:  Biomatik Corporation EKU12021
Encompass_Preferred
Human Kruppel Like Factor 15 (KLF15) CLIA Kit for detecting KLF15 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UIH9. Gene ID: 28999. Target Synonyms: KKLF; Kidney-Enriched Kruppel-Like Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3403