Learn More
Biomatik Corporation Acetylcarnitine (ALCAR) CLIA Kit
Supplier:  Biomatik Corporation EKU12097
Encompass_Preferred
Acetylcarnitine (ALCAR) CLIA Kit for detecting ALCAR in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 19.5-5,000ng/ml. Sensitivity: 7.7ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: Acetyl-L-carnitine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3415