Learn More
Biomatik Corporation Human Suppression Of Tumorigenicity 20 (ST20) ELISA Kit
Supplier:  Biomatik Corporation EKU12105
Encompass_Preferred
Human Suppression Of Tumorigenicity 20 (ST20) ELISA Kit for detecting ST20 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9HBF5. Target Synonyms: HCCS1; Human Cervical Cancer Suppressor Gene 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3418