Learn More
Biomatik Corporation Mouse Interferon Gamma (IFNg) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12111
Encompass_Preferred
Mouse Interferon Gamma (IFNg) Anti-antigen ELISA Kit for detecting IFNg in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.19ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P01580. Gene ID: 15978. Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3421