Learn More
Biomatik Corporation Human Glycated Albumin (GA) ELISA Kit
Supplier:  Biomatik Corporation EKU12124
Encompass_Preferred
Human Glycated Albumin (GA) ELISA Kit for detecting GA in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 246.9-20,000ng/ml. Sensitivity: 88.2ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3426