Learn More
Biomatik Corporation Mouse Superoxide Dismutase 2, Mitochondrial (SOD2) ELISA Kit
Supplier:  Biomatik Corporation EKU12165
Encompass_Preferred
Mouse Superoxide Dismutase 2, Mitochondrial (SOD2) ELISA Kit for detecting SOD2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09671. Gene ID: 20656. Target Synonyms: IPO-B; MNSOD; Mn-SOD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3439