Learn More
Biomatik Corporation Mouse Neurotensin (NT) ELISA Kit
Supplier:  Biomatik Corporation EKU12167
Encompass_Preferred
Mouse Neurotensin (NT) ELISA Kit for detecting NT in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 37.0-3,000pg/ml. Sensitivity: 14.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9D3P9. Gene ID: 67405. Target Synonyms: NTS; NTS1; NmN-125; NN; Tail peptide Neuromedin N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3441