Learn More
Biomatik Corporation Mouse Cortistatin (CORT) ELISA Kit
Supplier:  Biomatik Corporation EKU12197
Encompass_Preferred
Mouse Cortistatin (CORT) ELISA Kit for detecting CORT in serum, plasma, urine, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000ng/ml. Sensitivity: 5.13ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P56469. Gene ID: 12854. Target Synonyms: CST14; CST17; CST29. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3451