Learn More
Biomatik Corporation Human Myosin Heavy Chain 7B, Cardiac Muscle Beta (MYH7B) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12259
Encompass_Preferred
Human Myosin Heavy Chain 7B, Cardiac Muscle Beta (MYH7B) Anti-antigen ELISA Kit for detecting MYH7B in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: A7E2Y1. Gene ID: 57644. Target Synonyms: MYH14; MHC14; Antigen MLAA-21; Myosin cardiac muscle beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3467