Learn More
Biomatik Corporation Mouse Mab21 Domain Containing Protein 1 (MB21D1) ELISA Kit
Supplier:  Biomatik Corporation EKU12269
Encompass_Preferred
Mouse Mab21 Domain Containing Protein 1 (MB21D1) ELISA Kit for detecting MB21D1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8C6L5. Gene ID: 214763. Target Synonyms: C6orf150; cGAS; Cyclic GMP-AMP synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3471