Learn More
Biomatik Corporation Human Interleukin 4 (IL4) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12279
Encompass_Preferred
Human Interleukin 4 (IL4) High-sensitive ELISA Kit for detecting IL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05112. Gene ID: 3565. Target Synonyms: BSF1; BCGF1 ;B Cell Stimulatory Factor 1; Lymphocyte stimulatory factor 1; B Cell Growth Factor; Binetrakin; Pitrakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3476