Learn More
Biomatik Corporation Dog Leukemia Inhibitory Factor (LIF) ELISA Kit
Supplier:  Biomatik Corporation EKU12298
Encompass_Preferred
Dog Leukemia Inhibitory Factor (LIF) ELISA Kit for detecting LIF in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: J9NRL6. Target Synonyms: CDF; D-FACTOR; HILDA; MLPLI; Cholinergic Differentiation Factor; Differentiation-Stimulating Factor; Melanoma-Derived LPL Inhibitor; Emfilermin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3484