Learn More
Biomatik Corporation Uric Acid (UA) ELISA Kit
Supplier:  Biomatik Corporation EKU12315
Encompass_Preferred
Uric Acid (UA) ELISA Kit for detecting UA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 617.3-50,000ng/ml. Sensitivity: 243.1ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Urate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3488