Learn More
Biomatik Corporation Human Sequestosome 1 (SQSTM1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12327
Encompass_Preferred
Human Sequestosome 1 (SQSTM1) Mini sample ELISA Kit for detecting SQSTM1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13501. Gene ID: 8878. Target Synonyms: OSIL; PDB3; ZIP3; p62; p62B; p60; A170; Paget Disease Of Bone 3; Oxidative Stress Induced Like; EBI3-associated protein of 60 kDa; Ubiquitin-binding protein p62. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3491