Learn More
Biomatik Corporation Mouse Alkaline Phosphatase, Intestinal (ALPI) ELISA Kit
Supplier:  Biomatik Corporation EKU12348
Encompass_Preferred
Mouse Alkaline Phosphatase, Intestinal (ALPI) ELISA Kit for detecting ALPI in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24822. Target Synonyms: CIP; IAP; Calf Intestinal Alkaline Phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3496