Learn More
Biomatik Corporation Human Forkhead Box Protein A1 (FOXA1) ELISA Kit
Supplier:  Biomatik Corporation EKU12369
Encompass_Preferred
Human Forkhead Box Protein A1 (FOXA1) ELISA Kit for detecting FOXA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55317. Gene ID: 3169. Target Synonyms: FOX-A1; HNF3A; Hepatocyte Nuclear Factor 3,alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3502