Learn More
Biomatik Corporation Rat Gamma-Glutamyltransferase 1 (gGT1) ELISA Kit
Supplier:  Biomatik Corporation EKU12385
Encompass_Preferred
Rat Gamma-Glutamyltransferase 1 (gGT1) ELISA Kit for detecting gGT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.57ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07314. Gene ID: 116568. Target Synonyms: CD224; G-GT1; GTG; Gamma-glutamyltranspeptidase 1; Glutathione hydrolase 1; Leukotriene-C4 hydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3509