Learn More
Biomatik Corporation Rat Myosin IC (MYO1C) ELISA Kit
Supplier:  Biomatik Corporation EKU12394
Encompass_Preferred
Rat Myosin IC (MYO1C) ELISA Kit for detecting MYO1C in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63355. Gene ID: 65261. Target Synonyms: MMI-beta; MMIb; NMI; myr2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3512