Learn More
Biomatik Corporation Human C-type mannose receptor 2 (MRC2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33328
Encompass_Preferred
The Human C-type mannose receptor 2 (MRC2) ELISA Kit allows for quantitative determination of mannose receptor, C type 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9UBG0. Restriction: For Research Use Only.
Catalog No. 50-149-7816