Learn More
Biomatik Corporation Human C-Type Lectin Domain Family 4, Member A (CLEC4A) ELISA Kit
Supplier:  Biomatik Corporation EKU12430
Encompass_Preferred
Human C-Type Lectin Domain Family 4, Member A (CLEC4A) ELISA Kit for detecting CLEC4A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UMR7. Gene ID: 50856. Target Synonyms: CLECSF6; DCIR; DDB27; HDCGC13P; LLIR; C-type lectin DDB27; C-type lectin superfamily member 6; Dendritic cell immunoreceptor; Lectin-like immunoreceptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3520