Learn More
Biomatik Corporation Human Major Histocompatibility Complex Class I C (MHCC) ELISA Kit
Supplier:  Biomatik Corporation EKU12460
Encompass_Preferred
Human Major Histocompatibility Complex Class I C (MHCC) ELISA Kit for detecting MHCC in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10321. Gene ID: 3107. Target Synonyms: MHC-C;; HLAC; HLA-JY3; HLC-C; HLA-C; PSORS1 Leukocyte Antigen C; HLA class I histocompatibility antigen, Cw-7 alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3525