Learn More
Biomatik Corporation Human Hepcidin (Hepc) CLIA Kit
Supplier:  Biomatik Corporation EKU12467
Encompass_Preferred
Human Hepcidin (Hepc) CLIA Kit for detecting Hepc in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-20,000pg/ml. Sensitivity: 27.74pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P81172. Gene ID: 57817. Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3527