Learn More
Biomatik Corporation Human Apolipoprotein A1 (APOA1) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12511
Encompass_Preferred
Human Apolipoprotein A1 (APOA1) Anti-antigen ELISA Kit for detecting APOA1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P02647. Gene ID: 335. Target Synonyms: Apo-A1; ApoA-1 Milano; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3539