Learn More
Biomatik Corporation Canine Folic acid,FA ELISA Kit
Supplier:  Biomatik Corporation EKC31027
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Canis lupus familiaris (Dog) (Canis familiaris). Detection Range: 0.195 ng/mL-50 ng/mL. Sensitivity: 0.195 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3554