Learn More
Biomatik Corporation Dog Vitamin A, Va Elisa Kit
Supplier:  Biomatik Corporation EKC31047
Encompass_Preferred
Dog Vitamin A, Va Elisa Kit for detecting Vitamin A, Va in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-50ng/ml. Sensitivity: 2.336ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3556