Learn More
Biomatik Corporation Human C-C Motif Chemokine 16 (Ccl16) Elisa Kit
Supplier:  Biomatik Corporation EKC31189
Encompass_Preferred
Human C-C Motif Chemokine 16 (Ccl16) Elisa Kit for detecting Chemokine (C-C Motif) Ligand 16 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 39.06-2500pg/ml. Sensitivity: 9.77pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O15467; CCL16. Target Synonyms: C-C motif chemokine 16; CCL16; CCL16_HUMAN; Chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; HCC4; IL-10-inducible chemokine; IL10 inducible chemokine; IL10 inducible chemokine; ILINCK; LCC-1; LCC1; Liver CC chemokine 1 precursor; Liver expressed chemokine; Liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; Monotactin 1; Monotactin-1; MTN-1; Mtn1; NCC-4; NCC4; New CC chemokine 4; SCYA16; SCYL4; Small inducible cytokine A16 precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3572