Learn More
Biomatik Corporation Human Lymphatic Vessel Endothelial Hyaluronic Acid Receptor 1 (Lyve1) Elisa Kit
Supplier:  Biomatik Corporation EKC31276
Encompass_Preferred
Human Lymphatic Vessel Endothelial Hyaluronic Acid Receptor 1 (Lyve1) Elisa Kit for detecting Lymphatic Vessel Endothelial Hyaluronan Receptor 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 19.5-1250pg/ml. Sensitivity: 4.88pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y5Y7; LYVE1. Target Synonyms: Cell surface retention sequence-binding protein 1; CRSBP 1; CRSBP-1; CRSBP1; extracellular link domain containing 1; extracellular link domain-containing 1; Extracellular link domain-containing protein 1; HAR; Hyaluronic acid receptor; Lymphatic endothelium specific hyaluronan receptor; lymphatic vessel endothelial hyaluronan receptor 1; Lymphatic vessel endothelial hyaluronic acid receptor 1; LYVE 1; LYVE-1; LYVE1; LYVE1_HUMAN; XLKD1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3579