Learn More
Biomatik Corporation Human N-Terminal Pro-Brain Natriuretic Peptide, Nt-Probnp Elisa Kit
Supplier:  Biomatik Corporation EKC31297
Encompass_Preferred
Human N-Terminal Pro-Brain Natriuretic Peptide, Nt-Probnp Elisa Kit for detecting N-Terminal Pro-Brain Natriuretic Peptide, Nt-Probnp in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.216ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3582