Learn More
Biomatik Corporation Mouse Dickkopf-related protein 1(DKK1) ELISA kit
Supplier:  Biomatik Corporation EKC31391
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78.125 pg/mL-5000 pg/mL. Sensitivity: 13.127 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O54908. Target Synonyms: DKK-1, SK, dickkopf homolog 1|dickkopf related protein-1|dickkopf-1 like. Restriction: For Research Use Only.
Catalog No. 50-229-3598