Learn More
Biomatik Corporation Mouse growth arrest-specific gene-6(gas-6)ELISA Kit
Supplier:  Biomatik Corporation EKC31400
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 17.086 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q61592. Target Synonyms: AXLLG, AXSF, DKFZp666G247, FLJ34709, AXL receptor tyrosine kinase ligand|AXL stimulatory factor. Restriction: For Research Use Only.
Catalog No. 50-229-3601