Learn More
Biomatik Corporation Mouse Pentraxin-related protein PTX3(PTX3) ELISA kit
Supplier:  Biomatik Corporation EKC31416
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.313 ng/mL-20 ng/mL. Sensitivity: 0.210 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P48759. Target Synonyms: TNFAIP5, TSG-14, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, alpha-induced protein 5|pentaxin-related protein PTX3|pentraxin 3|pentraxin-3|tumor necrosis factor, alph. Restriction: For Research Use Only.
Catalog No. 50-229-3605